missing translation for 'onlineSavingsMsg'
Learn More

3-Cyanophenyl isocyanate, 97%

Product Code. 10073590 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10073590 5 g Glass bottle 5g
10117763 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10073590 Supplier Thermo Scientific Acros Supplier No. 314180050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 16413-26-6 | C8H4N2O | 144.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16413-26-6
Molecular Formula C8H4N2O
Molecular Weight (g/mol) 144.13
MDL Number MFCD00013856
InChI Key NZHPVPMRNASEQK-UHFFFAOYSA-N
Synonym 3-cyanophenyl isocyanate, 3-cyanophenylisocyanate, benzonitrile, 3-isocyanato, 3-isocyanatobenzenecarbonitrile, m-cyanophenyl isocyanate, m-cyanophenylisocyanate, pubchem5028, 3-isocyanato-benzonitrile, acmc-1c7a7, paragos 331038
PubChem CID 2733315
IUPAC Name 3-isocyanatobenzonitrile
SMILES C1=CC(=CC(=C1)N=C=O)C#N

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 49°C to 54°C
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula CNC6H4NCO
Quantity 5 g
Formula Weight 144.13
Percent Purity 97%
Physical Form Low Melting Solid
Chemical Name or Material 3-Cyanophenyl isocyanate, 97%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.