missing translation for 'onlineSavingsMsg'
Learn More

3-(Cyanomethyl)benzeneboronic acid pinacol ester, 96%, Thermo Scientific™

Product Code. 15439955
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15439955 250 mg 250mg
15419955 1 g 1g
15429955 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15439955 Supplier Thermo Scientific Alfa Aesar Supplier No. H52618.MD
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
3-(Cyanomethyl)benzeneboronic acid pinacol ester is used as pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Stable under recommended storage conditions. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 396131-82-1
Molecular Formula C14H18BNO2
Molecular Weight (g/mol) 243.11
MDL Number MFCD02179458
InChI Key LIWPKYSGWSFPEE-UHFFFAOYSA-N
Synonym 2-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl acetonitrile, 3-cyanomethyl phenylboronic acid pinacol ester, 2-3-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl acetonitrile, 3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl acetonitrile, pubchem7790, 3-cyanomethylphenylboronic acid, pinacol ester, 3-cyanomethylphenyl boronic acid,pinacol ester, 3-cyanomethyl phenylboronic acid, pinacol ester
PubChem CID 2773343
IUPAC Name 2-[3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]acetonitrile
SMILES CC1(C)OB(OC1(C)C)C1=CC(CC#N)=CC=C1

Specifications

Melting Point 44°C to 48°C
Quantity 250 mg
Solubility Information Slightly soluble in water.
Formula Weight 243.11
Percent Purity 96%
Chemical Name or Material 3-(Cyanomethyl)benzeneboronic acid pinacol ester
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.