missing translation for 'onlineSavingsMsg'
Learn More

3-Cyanocoumarin, 97%, Thermo Scientific™

Product Code. 15442172 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15442172 1 g 1g
15452172 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15442172 Supplier Thermo Scientific Acros Supplier No. 456590010
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 15119-34-3
Molecular Formula C10H5NO2
Molecular Weight (g/mol) 171.16
MDL Number MFCD00115699
InChI Key QKJALQPLNMEDAV-UHFFFAOYSA-N
Synonym 3-cyanocoumarin, 2-oxo-2h-chromene-3-carbonitrile, coumarin-3-carbonitrile, 2h-1-benzopyran-3-carbonitrile, 2-oxo, 2-oxo-2h-1-benzopyran-3-carbonitrile, coumarin, 3-cyano-7ci,8ci, 3-cyano-coumarin, pubchem8675, acmc-1cfdq
PubChem CID 203763
IUPAC Name 2-oxochromene-3-carbonitrile
SMILES O=C1OC2=CC=CC=C2C=C1C#N

Specifications

Melting Point 184.0°C to 188.0°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 171.15
Percent Purity 97%
Chemical Name or Material 3-Cyanocoumarin

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.