missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Cyanobenzoic Acid, TRC
Click to view available options
Quantity:
2.5 g
Unit Size:
2.5g
Specifications
Specifications
| Chemical Name or Material | 3-Cyanobenzoic Acid |
| CAS | 1877-72-1 |
| Molecular Formula | C8H5NO2 |
| Quantity | 2.5 g |
| Synonym | 3-Carboxybenzonitrile, m-Carboxybenzonitrile, m-Cyanobenzoic Acid |
| InChI Formula | InChI=1S/C8H5NO2/c9-5-6-2-1-3-7(4-6)8(10)11/h1-4H,(H,10,11) |
| SMILES | OC(=O)c1cccc(c1)C#N |
| IUPAC Name | 3-cyanobenzoic acid |
| Molecular Weight (g/mol) | 147.13 |
| Formula Weight | 147.032 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction