missing translation for 'onlineSavingsMsg'
Learn More

3-Cyanobenzoic Acid Methyl Ester, TRC

Product Code. 30358294
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30358294 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30358294 Supplier Toronto Research Chemicals Supplier No. TRCC9623151G
 Restricted Item
Estimated Shipment: 17-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 3-Cyanobenzoic Acid Methyl Ester
CAS 13531-48-1
Molecular Formula C9H7NO2
Quantity 1 g
Synonym Methyl 3-Cyanobenzoate, Methyl m-Cyanobenzoate, m-Cyanobenzoic Acid Methyl Ester, 3-(Methoxycarbonyl)benzonitrile
InChI Formula InChI=1S/C9H7NO2/c1-12-9(11)8-4-2-3-7(5-8)6-10/h2-5H,1H3
SMILES COC(=O)c1cccc(c1)C#N
IUPAC Name methyl 3-cyanobenzoate
Molecular Weight (g/mol) 161.16
Formula Weight 161.0477
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.