missing translation for 'onlineSavingsMsg'
Learn More

3-Cyanobenzoic acid, 98+%

Product Code. 11490308
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11490308 100 g 100g
11470308 5 g 5g
11480308 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11490308 Supplier Thermo Scientific Alfa Aesar Supplier No. B23836.22
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1877-72-1 | C8H5NO2 | 147.133 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1877-72-1
Molecular Formula C8H5NO2
Molecular Weight (g/mol) 147.133
MDL Number MFCD00002486
InChI Key GYLKKXHEIIFTJH-UHFFFAOYSA-N
Synonym m-cyanobenzoic acid, benzoic acid, 3-cyano, 3-carboxybenzonitrile, 3-cyano-benzoic acid, 3-cyanobenzoicacid, benzoic acid, m-cyano, unii-d2gc692d78, 3-cyano-benzoicacid, m-carboxybenzonitrile, 3-cyano benzoic acid
PubChem CID 15875
IUPAC Name 3-cyanobenzoic acid
SMILES C1=CC(=CC(=C1)C(=O)O)C#N

Specifications

Melting Point 219°C to 224°C
Quantity 100 g
Beilstein 1862566
Formula Weight 147.13
Percent Purity ≥98%
Chemical Name or Material 3-Cyanobenzoic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.