missing translation for 'onlineSavingsMsg'
Learn More

3-Cyanobenzenesulfonyl chloride, 97%

Product Code. 11348996
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11348996 5 g 5g
11338996 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11348996 Supplier Thermo Scientific Alfa Aesar Supplier No. L20169.06
 Restricted Item
Estimated Shipment: 03-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 56542-67-7 | C7H4ClNO2S | 201.624 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 56542-67-7
Molecular Formula C7H4ClNO2S
Molecular Weight (g/mol) 201.624
MDL Number MFCD00203480
InChI Key BHNRGBRMCNHNQD-UHFFFAOYSA-N
Synonym 3-cyanobenzene-1-sulfonyl chloride, 3-cyanobenzene-1-sulfonylchloride, 3-cyano-benzenesulfonyl chloride, 3-cyanobenzenesulphonyl chloride, benzenesulfonyl chloride, 3-cyano, pubchem5108, acmc-1au46, 3-cyanobenzenesulfonylchloride, ksc269e7j, akos bb-9463
PubChem CID 2801360
IUPAC Name 3-cyanobenzenesulfonyl chloride
SMILES C1=CC(=CC(=C1)S(=O)(=O)Cl)C#N

Specifications

Melting Point 47°C to 52°C
Boiling Point 114°C to 116°C (0.2 mmHg)
Flash Point >110°C (230°F)
Quantity 5 g
UN Number UN3261
Beilstein 2832925
Sensitivity Moisture sensitive
Formula Weight 201.63
Percent Purity 97%
Chemical Name or Material 3-Cyanobenzenesulfonyl chloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.