missing translation for 'onlineSavingsMsg'
Learn More

3-Cyanobenzene-1-sulfonyl chloride, 97%, Thermo Scientific™

Product Code. 10541703
Change view
Click to view available options
Quantity:
10 g
Packaging:
Amber Glass Bottle
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10541703 10 g Amber Glass Bottle 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10541703 Supplier Thermo Scientific Maybridge Supplier No. TL00666EA
 Restricted Item
Estimated Shipment: 04-11-2026
Add to basket
This item is not returnable. View return policy

Specifications

CAS 56542-67-7
CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Molecular Formula C7H4ClNO2S
Linear Formula CNC6H4SO2Cl
MDL Number MFCD00203480
Quantity 10 g
Synonym 3-cyanobenzene-1-sulfonyl chloride, 3-cyanobenzene-1-sulfonylchloride, 3-cyano-benzenesulfonyl chloride, 3-cyanobenzenesulphonyl chloride, benzenesulfonyl chloride, 3-cyano, pubchem5108, acmc-1au46, 3-cyanobenzenesulfonylchloride, ksc269e7j, akos bb-9463
InChI Key BHNRGBRMCNHNQD-UHFFFAOYSA-N
SMILES C1=CC(=CC(=C1)S(=O)(=O)Cl)C#N
IUPAC Name 3-cyanobenzenesulfonyl chloride
Molecular Weight (g/mol) 201.624
PubChem CID 2801360
Formula Weight 201.63
Percent Purity 97%
Chemical Name or Material 3-Cyanobenzene-1-sulfonyl chloride
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.