missing translation for 'onlineSavingsMsg'
Learn More

3-Cyano-5-fluorophenylboronic acid, 97%, Thermo Scientific™

Product Code. 10227657 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
Packaging:
Glass bottle
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10227657 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10227657 Supplier Thermo Scientific Acros Supplier No. 430460010
 Restricted Item
Estimated Shipment: 03-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 304858-67-1
CAS Min % 96.0
CAS Max % 100.0
Color White
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Molecular Formula C7H5BFNO2
MDL Number MFCD09263608
Quantity 1 g
Synonym 3-cyano-5-fluorophenyl boronic acid, 3-cyano-5-fluorobenzeneboronic acid, 3-borono-5-fluorobenzonitrile, boronic acid, 3-cyano-5-fluorophenyl-9ci, boronic acid, 3-cyano-5-fluorophenyl, pubchem17351, acmc-1csdr, ksc495g6d, 3-cyano-5-fluorophenylboronicacid
InChI Key DLYWCECHXBOCAS-UHFFFAOYSA-N
SMILES OB(O)C1=CC(=CC(F)=C1)C#N
IUPAC Name (3-cyano-5-fluorophenyl)boronic acid
Molecular Weight (g/mol) 164.93
PubChem CID 23435605
Formula Weight 164.93
Percent Purity 97%
Physical Form Crystalline Solid
Chemical Name or Material 3-Cyano-5-fluorophenylboronic acid
Show More Show Less
compliance-icons
Product Identifier
  • 3-Cyano-5-fluorophenylboronic acid
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.