missing translation for 'onlineSavingsMsg'
Learn More

3-Coumaranone, 97%

Product Code. 11409522
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11409522 1 g 1g
11419522 5 g 5g
11429522 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11409522 Supplier Thermo Scientific Alfa Aesar Supplier No. A10202.03
 Restricted Item
Estimated Shipment: 07-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 7169-34-8 | C8H6O2 | 134.13 g/mol

3-Coumaranone is used in the aurones, [2-benzylidenebenzofuran-3(2H)-ones] by reacting with appropriate aldehyde. It is involved in the condensation reaction with aldehydes in the presence of morpholine acetate to prepare substituted 2-(arylidene)benzofuran-3(ZH)-ones. In Horner-Wadsworth-Emmons reaction, it reacts with diethyl¬ cyanomethyl¬phosphonate to give 3-(cyanomethyl)benzofurans, which is an intermediate in the synthesis of antihyperglycemic agents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Coumaranone is used in the aurones, [2-benzylidenebenzofuran-3(2H)-ones] by reacting with appropriate aldehyde. It is involved in the condensation reaction with aldehydes in the presence of morpholine acetate to prepare substituted 2-(arylidene)benzofuran-3(ZH)-ones. In Horner-Wadsworth-Emmons reaction, it reacts with diethyl¬ cyanomethyl¬phosphonate to give 3-(cyanomethyl)benzofurans, which is an intermediate in the synthesis of antihyperglycemic agents.

Solubility
Slightly miscible with water.

Notes
Light sensitive. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7169-34-8
Molecular Formula C8H6O2
Molecular Weight (g/mol) 134.13
MDL Number MFCD00051810
InChI Key MGKPCLNUSDGXGT-UHFFFAOYSA-N
Synonym benzofuran-3 2h-one, 3-coumaranone, 3 2h-benzofuranone, 1-benzofuran-3 2h-one, coumaranone, benzofuran-3-one, coumaran-3-one, 2,3-dihydrobenzo b furan-3-one, 2,3-dihydro-1-benzofuran-3-one, 2h-benzofuran-3-one
PubChem CID 23556
IUPAC Name 1-benzofuran-3-one
SMILES O=C1COC2=CC=CC=C12

Specifications

Melting Point 97°C to 102°C
Quantity 1 g
Beilstein 115296
Sensitivity Light sensitive
Solubility Information Slightly miscible with water.
Formula Weight 134.13
Percent Purity 97%
Chemical Name or Material 3-Coumaranone

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.