missing translation for 'onlineSavingsMsg'
Learn More

3-Chlorothiophene, 98%, may contain up to 2% DMF

Product Code. 11455297
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11455297 5 g 5g
11465297 25 g 25g
11475297 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11455297 Supplier Thermo Scientific Alfa Aesar Supplier No. B22669.06
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 17249-80-8 | C4H3ClS | 118.578 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 17249-80-8
Molecular Formula C4H3ClS
Molecular Weight (g/mol) 118.578
MDL Number MFCD00043887
InChI Key QUBJDMPBDURTJT-UHFFFAOYSA-N
Synonym thiophene, 3-chloro, 3-chloro thiophene, 3-thienyl chloride, 3-chlorthiophen, 3-chloro-thiophene, 3-chloranylthiophene, pubchem5477, acmc-1cdmv, 3-chlorothiophene, ksc175g2h
PubChem CID 87017
IUPAC Name 3-chlorothiophene
SMILES C1=CSC=C1Cl

Specifications

Density 1.276
Boiling Point 137°C to 139°C
Flash Point 37°C (98°F)
Odor Unpleasant
Refractive Index 1.552
Quantity 5 g
UN Number UN1992
Beilstein 105255
Formula Weight 118.59
Percent Purity 98%
Assay may contain up to 2% DMF
Chemical Name or Material 3-Chlorothiophene
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.