missing translation for 'onlineSavingsMsg'
Learn More

3-chlorothiophene-2-carbaldehyde, 90%, Thermo Scientific™

Product Code. 10143553
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10143553 1 g Amber Glass Bottle 1g
10412304 250 mg Amber Glass Bottle 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10143553 Supplier Thermo Scientific Maybridge Supplier No. SEW06567DA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 67482-48-8
Molecular Formula C5H3ClOS
Molecular Weight (g/mol) 146.588
MDL Number MFCD01764973
InChI Key PJOJWMNHMJFFCR-UHFFFAOYSA-N
Synonym 3-chlorothiophene-2-carboxaldehyde, 3-chloro-2-thiophenecarboxaldehyde, 2-thiophenecarboxaldehyde, 3-chloro, pubchem21836, acmc-1b7xg, 2-formyl-3-chlorothiophene, chlorothiophen-2-carbaldehyde, ksc494c6j, 3-chloro-2-formylthiohene, 3-chloro-thiophene-2-carbaldehyde
PubChem CID 2735825
IUPAC Name 3-chlorothiophene-2-carbaldehyde
SMILES C1=CSC(=C1Cl)C=O

Specifications

CAS Min % 90.0
Assay Percent Range 90%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 146.6
Percent Purity 90%
Chemical Name or Material 3-chlorothiophene-2-carbaldehyde
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.