missing translation for 'onlineSavingsMsg'
Learn More

3-Chlorostyrene, 98%, stab. with 0.1% 4-tert-butylcatechol

Product Code. 11420268
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11420268 5 g 5g
11430268 25 g 25g
11440268 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11420268 Supplier Thermo Scientific Alfa Aesar Supplier No. B23806.06
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 2039-85-2 | C8H7Cl | 138.594 g/mol

It has been identified as a potential candidate for an electron resist to be used in direct device fabrication.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It has been identified as a potential candidate for an electron resist to be used in direct device fabrication.

Solubility
Not miscible or difficult to mix in water. Soluble in ether, and ethanol

Notes
Store away from heat and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Keep away from ignition sources.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2039-85-2
Molecular Formula C8H7Cl
Molecular Weight (g/mol) 138.594
MDL Number MFCD00000598
InChI Key BOVQCIDBZXNFEJ-UHFFFAOYSA-N
Synonym 3-chlorostyrene, 1-chloro-3-vinylbenzene, m-chlorostyrene, benzene, 1-chloro-3-ethenyl, styrene, m-chloro, unii-11k8g759hc, 1-chloro-3-vinyl-benzene, styrene, 3-chloro, meta-chlorostyrene, zlchem 318
PubChem CID 14905
IUPAC Name 1-chloro-3-ethenylbenzene
SMILES C=CC1=CC(=CC=C1)Cl

Specifications

Density 1.09
Boiling Point 188°C to 189°C
Flash Point 67°C (153°F)
Odor Characteristic
Refractive Index 1.562
Quantity 5 g
Beilstein 2038489
Solubility Information Not miscible or difficult to mix in water. Soluble in ether,and ethanol
Formula Weight 138.6
Percent Purity 98%
Chemical Name or Material 3-Chlorostyrene, stabilized with 0.1% 4-tert-butylcatechol
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.