missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Chloropyridine-d4, CDN
Click to view available options
Quantity:
0.1 g
0.25 g
Unit Size:
0.1g
0.25g
Specifications
Specifications
| Chemical Name or Material | 3-Chloropyridine-D4 |
| CAS | 1001003-95-7 |
| Molecular Formula | C5 D4 Cl N |
| Quantity | 0.1 g |
| Synonym | Pyridine-2,3,4,6-d4, 5-chloro- (ACI), 5-Chloropyridine-2,3,4,6-d4 (ACI), 3-Chloropyridine-d4, 3-Chloropyridine-D4, 3-Chloropyridine-2,4,5,6-d4 |
| InChI Formula | InChI=1 S/C5H4ClN/c6-5-2-1-3-7-4-5/h1-4 H/i1D,2 D,3 D,4 D |
| SMILES | [2 H]c1nc([2 H])c(Cl)c([2 H])c1[2 H] |
| IUPAC Name | 3-chloro-2,4,5,6-tetradeuteriopyridine |
| Molecular Weight (g/mol) | 117.5696 |
| Formula Weight | 117.0283 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction