missing translation for 'onlineSavingsMsg'
Learn More

3-Chloropyrazine-2-carbonitrile, 95+%, Thermo Scientific™

Product Code. 10082444
Change view
Click to view available options
Quantity:
1 g
10 g
25 g
Packaging:
Amber Glass Bottle
Unit Size:
10g
1g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10082444 10 g Amber Glass Bottle 10g
10292442 1 g Amber Glass Bottle 1g
10527864 25 g Amber Glass Bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10082444 Supplier Thermo Scientific Maybridge Supplier No. KM04911EA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 55557-52-3
Molecular Formula C5H2ClN3
Molecular Weight (g/mol) 139.54
MDL Number MFCD00219653
InChI Key SDLFAEGTVBPHBK-UHFFFAOYSA-N
Synonym 2-chloro-3-cyanopyrazine, 3-chloro-2-pyrazinecarbonitrile, 2-chloro-3-cyannopyrazine, pyrazinecarbonitrile, 3-chloro, 3-chloro-2-cyanopyrazine, 3-chlorpyrazin-2-carbonitril, chlorocyanopyrazine, pubchem10180, acmc-209lnr, 2-cyano-3-chloropyrazine
PubChem CID 292465
IUPAC Name 3-chloropyrazine-2-carbonitrile
SMILES ClC1=NC=CN=C1C#N

Specifications

Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 10 g
Formula Weight 139.54
Percent Purity 97%
Chemical Name or Material 3-Chloropyrazine-2-carbonitrile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.