missing translation for 'onlineSavingsMsg'
Learn More

(3-chloropropyl)trimethoxysilane, 98+%

Product Code. 10187653 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
Packaging:
Glass bottle
Unit Size:
100mL
25mL
500mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10187653 500 mL Glass bottle 500mL
10051390 25 mL Glass bottle 25mL
10694992 100 mL Glass bottle 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10187653 Supplier Thermo Scientific Acros Supplier No. 137285000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 2530-87-2 | C6H15ClO3Si | 198.72 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2530-87-2
Molecular Formula C6H15ClO3Si
Molecular Weight (g/mol) 198.72
MDL Number MFCD00000997
InChI Key OXYZDRAJMHGSMW-UHFFFAOYSA-N
Synonym 3-chloropropyl trimethoxysilane, silane, 3-chloropropyl trimethoxy, 3-chloropropyltrimethyoxysilane, sila-ace s 620, cps-m, silane 3-chloropropyl tris methoxy, 3-trimethoxysilyl propyl chloride, unii-t21bnl1s7f, cptmo, 3-chloropropyl trimethoxysilan
PubChem CID 62449
IUPAC Name 3-chloropropyl(trimethoxy)silane
SMILES CO[Si](CCCCl)(OC)OC

Specifications

CAS Min % 98.0
CAS Max % 100.0
Color Colorless to Yellow
Density 1.0700g/mL
Boiling Point 195°C to 196°C (750.0 mmHg)
Flash Point 78°C
Infrared Spectrum Authentic
Assay Percent Range 98% min. (GC)
Packaging Glass bottle
Linear Formula (CH3O)3Si(CH2)3Cl
Refractive Index 1.4190 to 1.421
Quantity 500 mL
Specific Gravity 1.07
Solubility Information Solubility in water: soluble
Formula Weight 198.72
Percent Purity 98+%
Physical Form Liquid
Chemical Name or Material (3-Chloropropyl)trimethoxysilane
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.