missing translation for 'onlineSavingsMsg'
Learn More

(3-Chloropropyl)trimethoxysilane, 97%

Product Code. 11450707
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11450707 500 g 500g
11440707 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11450707 Supplier Thermo Scientific Alfa Aesar Supplier No. B22517.36
 Restricted Item
Estimated Shipment: 26-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 2530-87-2 | C6H15ClO3Si | 198.718 g/mol

(3-Chloropropyl)trimethoxysilane is a good coupling agent for epoxy resins to glass in laminates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(3-Chloropropyl)trimethoxysilane is a good coupling agent for epoxy resins to glass in laminates.

Solubility
Hydrolyzes in water.

Notes
Moisture sensitive. Store away from strong oxidizing agents, water/ moisture.
TRUSTED_SUSTAINABILITY

Identifiants chimiques

CAS 2530-87-2
Molecular Formula C6H15ClO3Si
Molecular Weight (g/mol) 198.718
MDL Number MFCD00000997
InChI Key OXYZDRAJMHGSMW-UHFFFAOYSA-N
Synonym 3-chloropropyl trimethoxysilane, silane, 3-chloropropyl trimethoxy, 3-chloropropyltrimethyoxysilane, sila-ace s 620, cps-m, silane 3-chloropropyl tris methoxy, 3-trimethoxysilyl propyl chloride, unii-t21bnl1s7f, cptmo, 3-chloropropyl trimethoxysilan
PubChem CID 62449
IUPAC Name 3-chloropropyl(trimethoxy)silane
SMILES CO[Si](CCCCl)(OC)OC

Spécification

Boiling Point 195°C to 197°C
Melting Point -50°C
Quantity 500 g
Flash Point 78°C (172°F)
Odor Recognizable
Refractive Index 1.42
Sensitivity Moisture sensitive
Density 1.077
Beilstein 969627
Solubility Information Hydrolyzes in water.
Formula Weight 198.72
Percent Purity 97%
Chemical Name or Material (3-Chloropropyl)trimethoxysilane
Afficher plus Afficher moins

RUO – Research Use Only

Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.