missing translation for 'onlineSavingsMsg'
Learn More

3-Chloropropyl isocyanate, 96%, Thermo Scientific™

Product Code. 10368790 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 10368790

Brand: Thermo Scientific Acros 314590050

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 13010-19-0
Molecular Formula C4H6ClNO
Molecular Weight (g/mol) 119.55
MDL Number MFCD00010384
InChI Key RQAVSDINDRNIKL-UHFFFAOYSA-N
Synonym 3-chloropropyl isocyanate, 3-chloropropylisocyanate, propane, 1-chloro-3-isocyanato, 1-chloro-3-isocyanato-propane, chloropropylisocyanate, chloropropyl isocyanate, 3-chloropropanisocyanate, chloro propyl isocyanate, acmc-20an5q, 3-chloropropyl-isocyanate
PubChem CID 166752
IUPAC Name 1-chloro-3-isocyanatopropane
SMILES ClCCCN=C=O

Specifications

CAS Min % 95.0
CAS Max % 100.0
Color Colorless
Density 1.1600g/mL
Boiling Point 152°C to 154°C
Flash Point 61°C
Infrared Spectrum Authentic
Assay Percent Range 95% min. (GC)
Packaging Glass bottle
Linear Formula Cl(CH2)3NCO
Refractive Index 1.4490 to 1.453
Quantity 5 g
Specific Gravity 1.16
Formula Weight 119.55
Percent Purity 96%
Physical Form Liquid
Chemical Name or Material 3-Chloropropyl isocyanate, 96%
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.