missing translation for 'onlineSavingsMsg'
Learn More

3-Chloropropyl acetate, 97+%

Product Code. 11491607
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11491607 25 g 25g
11401617 100 g 100g
11411617 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11491607 Supplier Thermo Scientific Alfa Aesar Supplier No. B25013.14
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 628-09-1 | C5H9ClO2 | 136.575 g/mol

3-Chloropropyl acetate is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agro chemicals, dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Chloropropyl acetate is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agro chemicals, dyestuff.

Solubility
Soluble in water (partly).

Notes
Store in a cool, dry place in tightly closed container. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 628-09-1
Molecular Formula C5H9ClO2
Molecular Weight (g/mol) 136.575
MDL Number MFCD00001000
InChI Key KPOHQIPNNIMWRL-UHFFFAOYSA-N
Synonym 1-propanol, 3-chloro-, acetate, 3-chloropropanol acetate, 3-chlorprop-1-ylacetat, gamma-chloropropyl acetate, 3-chlorprop-1-ylacetat german, acetic acid 3-chloro-propyl ester, 1-acetoxy-3-chloropropane, .gamma.-chloropropyl acetate, 3-chloro-propyl acetate, acetic acid 3-chloropropyl ester
PubChem CID 12334
IUPAC Name 3-chloropropyl acetate
SMILES CC(=O)OCCCCl

Specifications

Density 1.12
Boiling Point 70°C to 72°C (15 mmHg)
Flash Point 68°C (154°F)
Refractive Index 1.429
Quantity 25 g
Solubility Information Soluble in water (partly).
Formula Weight 136.58
Percent Purity 98%
Chemical Name or Material 3-Chloropropyl acetate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.