missing translation for 'onlineSavingsMsg'
Learn More

3-Chloropropiophenone, TRC

Product Code. 17992565
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17992565 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17992565 Supplier Toronto Research Chemicals Supplier No. TRCC37977010G
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

Denne vare kan ikke returneres. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Tekniske data

CAS 34841-35-5
Molecular Formula C9 H9 Cl O
Quantity 10 g
Synonym 3-Chloropropiophenone, 1-Propanone, 1-(3-chlorophenyl)- (9CI, ACI), 1-(3-Chlorophenyl)-1-propanone (ACI), Propiophenone, 3'-chloro- (6CI, 7CI), 3-Chlorophenyl ethyl ketone, 3'-Chloropropiophenone, m-Chloropropiophenone
InChI Formula InChI=1S/C9H9ClO/c1-2-9(11)7-4-3-5-8(10)6-7/h3-6H,2H2,1H3
SMILES CCC(=O)c1cccc(Cl)c1
IUPAC Name 1-(3-chlorophenyl)propan-1-one
Molecular Weight (g/mol) 168.6202
Formula Weight 168.0342
Chemical Name or Material 3-Chloropropiophenone
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.