missing translation for 'onlineSavingsMsg'
Learn More

3'-Chloropropiophenone, 98%

Product Code. 11444224
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11444224 25 g 25g
11454224 100 g 100g
11464224 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11444224 Supplier Thermo Scientific Alfa Aesar Supplier No. A12542.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 34841-35-5 | C9H9ClO | 168.62 g/mol

3'-Chloropropiophenone has been used in the preparation of 1-(3-chlorophenyl)-1-phenyl-1-propanol.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3′-Chloropropiophenone has been used in the preparation of 1-(3-chlorophenyl)-1-phenyl-1-propanol.

Solubility
Soluble in methanol.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 34841-35-5
Molecular Formula C9H9ClO
Molecular Weight (g/mol) 168.62
MDL Number MFCD00009925
InChI Key PQWGFUFROKIJBO-UHFFFAOYSA-N
Synonym 3'-chloropropiophenone, 1-3-chlorophenyl propan-1-one, m-chloropropiophenone, 3-chlorophenyl ethyl ketone, 1-propanone, 1-3-chlorophenyl, 1-3-chloro-phenyl-propan-1-one, m-chloro-propiophenone, pubchem3083, 3'-chloro-propiophenone, 3/'-chloropropiophenone
PubChem CID 587128
IUPAC Name 1-(3-chlorophenyl)propan-1-one
SMILES CCC(=O)C1=CC(=CC=C1)Cl

Specifications

Melting Point 45°C to 47°C
Boiling Point 123°C to 124°C (13 mmHg)
Flash Point >110°C (230°F)
Quantity 25 g
Solubility Information Soluble in methanol.
Formula Weight 168.62
Percent Purity 98%
Chemical Name or Material 3'-Chloropropiophenone

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.