missing translation for 'onlineSavingsMsg'
Learn More

3'-Chloropropiophenone, 98%

Product Code. 10114690 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
250 g
Packaging:
Glass bottle
Unit Size:
250g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10114690 250 g Glass bottle 250g
10287260 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10114690 Supplier Thermo Scientific Acros Supplier No. 276582500
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 34841-35-5 | C9H9ClO | 168.62 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 34841-35-5
Molecular Formula C9H9ClO
Molecular Weight (g/mol) 168.62
MDL Number MFCD00009925
InChI Key PQWGFUFROKIJBO-UHFFFAOYSA-N
Synonym 3'-chloropropiophenone, 1-3-chlorophenyl propan-1-one, m-chloropropiophenone, 3-chlorophenyl ethyl ketone, 1-propanone, 1-3-chlorophenyl, 1-3-chloro-phenyl-propan-1-one, m-chloro-propiophenone, pubchem3083, 3'-chloro-propiophenone, 3/'-chloropropiophenone
PubChem CID 587128
IUPAC Name 1-(3-chlorophenyl)propan-1-one
SMILES CCC(=O)C1=CC(=CC=C1)Cl

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 43°C to 47°C
Color White to Yellow
Boiling Point 124°C (14.0 mmHg)
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula ClC6H4COC2H5
Quantity 250 g
Beilstein 07, 1028
Solubility Information Solubility in water: soluble in hot water. Other solubilities: soluble in hot toluene and alcohol
Formula Weight 168.62
Percent Purity 98%
Physical Form Crystalline Solid
Chemical Name or Material 3'-Chloropropiophenone, 98%
Show More Show Less
compliance-icons
Product Identifier
  • 3'-Chloropropiophenone
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.