missing translation for 'onlineSavingsMsg'
Learn More

3-Chloropropiophenone, 96%

Product Code. 11435023
Change view
Click to view available options
Quantity:
10 g
50 g
100 g
Unit Size:
100g
10g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11435023 50 g 50g
11425023 10 g 10g
11309499 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11435023 Supplier Thermo Scientific Alfa Aesar Supplier No. A12290.18
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 936-59-4 | C9H9ClO | 168.62 g/mol

3-Chloropropiophenone was used in the asymmetric reduction of (S)-3-chloro-1-phenylpropanol using preheated Candida utilis cells immobilized in calcium alginate gel beads. It was also used in the synthesis of (R)-3-chloro-1-phenyl-1-propanol via asymmetric reduction using in-situ generated oxazaborolidine catalyst derived from (S)-α,α-diphenylprolinol. It is a precursor of phenyl vinyl ketone.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Chloropropiophenone was used in the asymmetric reduction of (S)-3-chloro-1-phenylpropanol using preheated Candida utilis cells immobilized in calcium alginate gel beads. It was also used in the synthesis of (R)-3-chloro-1-phenyl-1-propanol via asymmetric reduction using in-situ generated oxazaborolidine catalyst derived from (S)-α,α-diphenylprolinol. It is a precursor of phenyl vinyl ketone.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 936-59-4
Molecular Formula C9H9ClO
Molecular Weight (g/mol) 168.62
MDL Number MFCD00000990
InChI Key KTJRGPZVSKWRTJ-UHFFFAOYSA-N
Synonym 3-chloropropiophenone, 1-propanone, 3-chloro-1-phenyl, beta-chloropropiophenone, 2-chloroethyl phenyl ketone, 3-chloropropanophenone, 3-chloro-1-phenyl-1-propanone, propiophenone, 3-chloro, 3-chloro-1-phenyl-propan-1-one, .beta.-chloropropiophenone, .omega.-chloropropiophenone
PubChem CID 70295
IUPAC Name 3-chloro-1-phenylpropan-1-one
SMILES C1=CC=C(C=C1)C(=O)CCCl

Specifications

Melting Point 46°C to 50°C
Boiling Point 113°C to 115°C (4 mmHg)
Flash Point >110°C (230°F)
Quantity 50 g
Beilstein 2043580
Solubility Information Insoluble in water.
Formula Weight 168.62
Percent Purity 96%
Chemical Name or Material 3-Chloropropiophenone

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.