Learn More
3-Chloropropionitrile, 98%
Description
3-Chloropropionitrile is used in pharmaceutical and polymer synthesis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
3-Chloropropionitrile is used in pharmaceutical and polymer synthesis.
Solubility
Soluble in water. (4.5g/100mL, 25°C). Miscible in Ether, Alcohols, Benzene, Acetone.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible materials are Strong acids, Strong bases, Strong oxidizing agents, Strong reducing agents.
Specifications
Specifications
| Melting Point | -51°C |
| Density | 1.143 |
| Boiling Point | 174°C to 176°C |
| Flash Point | 75°C (167°F) |
| Refractive Index | 1.438 |
| Quantity | 100 g |
| UN Number | UN3276 |
| Beilstein | 1098454 |
| Merck Index | 14,2160 |
| Solubility Information | Soluble in water. (4.5g/100mL,25°C). Miscible in Ether,Alcohols,Benzene,Acetone. |
| Show More |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.