missing translation for 'onlineSavingsMsg'
Learn More

3-Chloropropionic Acid, TRC

Product Code. 30137523
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
Unit Size:
10g
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30137523 5 g 5g
30138746 10 g 10g
17973635 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30137523 Supplier Toronto Research Chemicals Supplier No. TRCC5880305G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 107-94-8
Molecular Formula C3H5ClO2
Molecular Weight (g/mol) 108.52
Synonym Propanoic acid, 3-chloro- (9CI, ACI), 3-Chloropropanoic acid (ACI), Propionic acid, 3-chloro- (7CI, 8CI), Propionic acid, β-chloro- (2CI), 3-Chloropropionic acid, NSC 174, NSC 2183, β-Chloropropionic acid, β-Monochloropropionic acid
IUPAC Name 3-chloropropanoic acid
SMILES C(CCl)C(=O)O

Specifications

Quantity 5 g
InChI Formula InChI=1S/C3H5ClO2/c4-2-1-3(5)6/h1-2H2,(H,5,6)
Formula Weight 107.9978
Chemical Name or Material 3-Chloropropionic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.