missing translation for 'onlineSavingsMsg'
Learn More

3-Chloropropane-1,2-diol, Mikromol™

Product Code. 18080150
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18080150 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18080150 Supplier Mikromol Supplier No. MM0027.22

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

3-Chloropropane-1,2-diol
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 3-Chloropropane-1,2-diol
CAS 96-24-2
Analyte or Component Names 3-Chloropropane-1,2-diol
Shipping Condition Room Temperature
Molecular Formula C3 H7 Cl O2
Certifications/Compliance ISO 17025
Synonym 3-monochloropropanediol, 3-MCPD, 3-Chloro-1,2-propanediol, (RS)-3-Chloro-1,2-propanediol, (RS)-α-Chlorohydrin, (±)-2,3-Dihydroxychloropropane, (±)-3-Chloro-1,2-propanediol, 1,2-Dihydroxy-3-chloropropane, 1-Chloro-1-deoxyglycerol, 1-Chloro-2,3-dihydroxypropane, 1-Chloro-2,3-propanediol, 3-Chloro-1,2-dihydroxypropane, 3-Chloro-1,2-propylene glycol, 3-Chloropropanediol, 3-Chloropropylene glycol, 3-Monochloro-1,2-propanediol, Chlorohydrin, Chloropropanediol, Ekorod A, Epibloc, Glycerin epichlorohydrin, Glycerin monochlorohydrin, Glycerin α-monochlorhydrin, Glycerol 3-chlorohydrin, Glycerol monochlorohydrin, Glycerol α-chlorohydrin, Glycerol α-monochlorohydrin, Glyceryl chloride, Glyceryl α-chlorohydrin, U 5897, dl-α-Chlorohydrin, α-Chlorohydrin, α-Monochlorohydrin
InChI Formula InChI=1S/C3H7ClO2/c4-1-3(6)2-5/h3,5-6H,1-2H2
SMILES OCC(O)CCl
IUPAC Name 3-chloropropane-1,2-diol
Molecular Weight (g/mol) 110.5395
Quantity 100 mg
Formula Weight 110.0135 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.