missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Chloropicolinic Acid, TRC
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
Specifications
Specifications
| Chemical Name or Material | 3-Chloropicolinic Acid |
| CAS | 57266-69-0 |
| Molecular Formula | C6 H4 Cl N O2 |
| Quantity | 250 mg |
| InChI Formula | InChI=1S/C6H4ClNO2/c7-4-2-1-3-8-5(4)6(9)10/h1-3H,(H,9,10) |
| SMILES | OC(=O)c1ncccc1Cl |
| IUPAC Name | 3-chloropyridine-2-carboxylic acid |
| Molecular Weight (g/mol) | 157.55 |
| Formula Weight | 156.9931 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction