missing translation for 'onlineSavingsMsg'
Learn More

3-Chlorophenylmagnesium bromide, 1M in MeTHF

Product Code. 15498037
Change view
Click to view available options
Quantity:
50 mL
100 mL
Unit Size:
100mL
50mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15498037 50 mL 50mL
15408047 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15498037 Supplier Thermo Scientific Alfa Aesar Supplier No. H51161.AD
 Restricted Item
Estimated Shipment: 18-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 36229-42-2 | C6H4BrClMg | 215.76 g/mol

3-Chlorophenylmagnesium bromide is involved in the synthesis of enantiopure 2-imidazolidinones by reacting with 1,1'-DiBoc-2,2'-Biaziridine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Chlorophenylmagnesium bromide is involved in the synthesis of enantiopure 2-imidazolidinones by reacting with 1,1′-DiBoc-2,2′-Biaziridine.

Notes
Air and moisture sensitive. Incompatible with strong oxidizing agents. Reacts with water.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 36229-42-2
Molecular Formula C6H4BrClMg
Molecular Weight (g/mol) 215.76
MDL Number MFCD00672003
InChI Key ZIYRUWPTUPFRQU-UHFFFAOYSA-M
Synonym 3-chlorophenylmagnesium bromide, 3-chlorophenyl magnesium bromide, magnesium,bromo 3-chlorophenyl, m-chlorophenylmagnesium bromide, m-chlorophenyl magnesium bromide, 3-chloro phenyl magnesium bromide, 3-chloro-phenyl magnesium bromide, ziyruwptupfrqu-uhfffaoysa-m, grignard reagent
PubChem CID 12252967
IUPAC Name magnesium;chlorobenzene;bromide
SMILES ClC1=CC([Mg]Br)=CC=C1

Specifications

Quantity 50 mL
UN Number UN2924
Formula Weight 215.76
Concentration or Composition (by Analyte or Components) 1M in MeTHF
Chemical Name or Material 3-Chlorophenylmagnesium bromide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.