missing translation for 'onlineSavingsMsg'
Learn More

3-Chlorophenylboronic Acid, TRC

Product Code. 30350219
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30350219 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30350219 Supplier Toronto Research Chemicals Supplier No. TRCC59651050MG
 Restricted Item
Estimated Shipment: 21-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 3-Chlorophenylboronic Acid
CAS 63503-60-6
Molecular Formula C6 H6 B Cl O2
Quantity 50 mg
Synonym B-(3-Chlorophenyl)boronic Acid, (3-Chlorophenyl)boronic Acid, (m-Chlorophenyl)boronic Acid, 3-Chlorobenzeneboronic Acid, m-Chlorobenzeneboronic Acid
InChI Formula InChI=1S/C6H6BClO2/c8-6-3-1-2-5(4-6)7(9)10/h1-4,9-10H
SMILES OB(O)c1cccc(Cl)c1
IUPAC Name (3-chlorophenyl)boronic acid
Molecular Weight (g/mol) 156.375
Formula Weight 156.015
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.