missing translation for 'onlineSavingsMsg'
Learn More

3-Chlorophenylboronic acid, 97%

Product Code. 10311584 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10311584 1 g Glass bottle 1g
10140082 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10311584 Supplier Thermo Scientific Acros Supplier No. 359000010
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 63503-60-6 | C6H6BClO2 | 156.37 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 63503-60-6
Molecular Formula C6H6BClO2
Molecular Weight (g/mol) 156.37
MDL Number MFCD00161354
InChI Key SDEAGACSNFSZCU-UHFFFAOYSA-N
Synonym 3-chlorophenyl boronic acid, 3-chlorobenzeneboronic acid, 3-chlorophenyl boranediol, 3-chlorophenylbornic acid, 3-chloro benzene boronic acid, m-chlorophenylboronic acid, 3-chloro-phenyl-boronic acid, boronic acid, 3-chlorophenyl, pubchem1785
PubChem CID 2734323
IUPAC Name (3-chlorophenyl)boronic acid
SMILES OB(O)C1=CC(Cl)=CC=C1

Specifications

Melting Point 185°C to 189°C
CAS Min % 96.0
Color Green-Yellow to White
CAS Max % 100.0
Quantity 1 g
Infrared Spectrum Authentic
Packaging Glass bottle
Physical Form Crystalline Powder or Needles
Assay Percent Range 96% min. (HPLC)
Formula Weight 156.38
Percent Purity 97%
Chemical Name or Material 3-Chlorophenylboronic acid
Show More Show Less
compliance-icons
Product Identifier
  • 3-Chlorophenylboronic acid
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.