missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Chlorophenylboronic acid, 97%, May contain varying amounts of anhydride, Thermo Scientific™
Click to view available options
Amber Glass Bottle; 1g
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
CAS | 63503-60-6 |
CAS Min % | 96.0 |
Color | Green-Yellow to White |
CAS Max % | 100.0 |
Assay Percent Range | 97% |
Molecular Formula | C6H6BClO2 |
MDL Number | MFCD00161354 |
Synonym | 3-chlorophenyl boronic acid, 3-chlorobenzeneboronic acid, 3-chlorophenyl boranediol, 3-chlorophenylbornic acid, 3-chloro benzene boronic acid, m-chlorophenylboronic acid, 3-chloro-phenyl-boronic acid, boronic acid, 3-chlorophenyl, pubchem1785 |
InChI Key | SDEAGACSNFSZCU-UHFFFAOYSA-N |
SMILES | OB(O)C1=CC(Cl)=CC=C1 |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'