missing translation for 'onlineSavingsMsg'
Learn More

3-Chlorophenylacetone, 94%

Product Code. 10456822 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10456822 1 g Glass bottle 1g
10304642 5 g Glass bottle 5g
11994851 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10456822 Supplier Thermo Scientific Acros Supplier No. 350690010
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 14123-60-5 | C9H9ClO | 168.62 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14123-60-5
Molecular Formula C9H9ClO
Molecular Weight (g/mol) 168.62
MDL Number MFCD00082872
InChI Key VCNYPJMEQHTAHS-UHFFFAOYSA-N
PubChem CID 2734097
IUPAC Name 1-(3-chlorophenyl)propan-2-one
SMILES CC(=O)CC1=CC=CC(Cl)=C1

Specifications

Color Yellow
Infrared Spectrum Authentic
Assay Percent Range 92% min. (GC)
Packaging Glass bottle
Refractive Index 1.5320 to 1.5360
Quantity 1 g
Beilstein 07,IV,691
Synonym 1-3-chlorophenyl propan-2-one, 3-chlorophenylacetone, 1-3-chlorophenyl acetone, 1-3-chlorophenyl-2-propanone, pubchem19805, 3-chlorophenyl-acetone, m-chlorophenyl acetone, 2-propanone,1-3-chlorophenyl, 1-3-chloro-phenyl-propan-2-one, 2-propanone, 1-3-chlorophenyl
Formula Weight 168.62
Percent Purity 94%
Physical Form Liquid
Chemical Name or Material 3-Chlorophenylacetone
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.