missing translation for 'onlineSavingsMsg'
Learn More

3-(Chloromethyl)pyridine Hydrochloride, TRC

Product Code. 30351873
Change view
Click to view available options
Quantity:
2.5 g
25 g
Unit Size:
2.5g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30351873 25 g 25g
30350119 2.5 g 2.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30351873 Supplier Toronto Research Chemicals Supplier No. TRCC36981525G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 3-(Chloromethyl)pyridine Hydrochloride
CAS 6959-48-4
Molecular Formula C6H7Cl2N
Quantity 25 g
Synonym (Pyridin-3-yl)methyl Chloride Hydrochloride, 3-(Chloromethyl)pyridine Monohydrochloride, 3-Picolyl Chloride Hydrochloride, 3-Pyridylmethyl Chloride Hydrochloride
InChI Formula InChI=1S/C6H6ClN.ClH/c7-4-6-2-1-3-8-5-6;/h1-3,5H,4H2;1H
SMILES Cl.ClCc1cccnc1
IUPAC Name 3-(chloromethyl)pyridine;hydrochloride
Molecular Weight (g/mol) 164.03
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.