missing translation for 'onlineSavingsMsg'
Learn More

3-(chloromethyl)benzamide, 97%, Thermo Scientific™

Product Code. 10440564
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10440564 5 g Amber Glass Bottle 5g
10714583 250 mg Amber Glass Bottle 250mg
10100702 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10440564 Supplier Thermo Scientific Maybridge Supplier No. S08130DE
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 135654-16-9
Molecular Formula C8H8ClNO
Molecular Weight (g/mol) 169.608
MDL Number MFCD00175948
InChI Key OOYATKQQGVPYQR-UHFFFAOYSA-N
Synonym 3-chloromethyl benzamide, benzamide,3-chloromethyl, benzamide, 3-chloromethyl-9ci, 3-chloromethyl-benzamide, 3-carbamoylbenzyl chloride, acmc-20a41t, 3-aminocarbonyl benzyl chloride, 3-carbamoylbenzyl chloride, 3-aminocarbonyl benzyl chloride
PubChem CID 737145
IUPAC Name 3-(chloromethyl)benzamide
SMILES C1=CC(=CC(=C1)CCl)C(=O)N

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Linear Formula CH2ClC6H4C(O)NH2
Quantity 5 g
Formula Weight 169.61
Percent Purity 97%
Chemical Name or Material 3-(chloromethyl)benzamide
compliance-icons
Product Identifier
  • 3-(Chloromethyl)benzamide
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.