missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-(Chloromethyl)-5-phenyl-1,2,4-oxadiazole, 97%, Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
5 g
Specifications
CAS Min % | 97.0 |
Assay Percent Range | 97% |
Packaging | Amber Glass Bottle |
Quantity | 250 mg |
Synonym | 3-chloromethyl-5-phenyl-1,2,4-oxadiazole, 3-chloromethyl-5-phenyl-1,2,4 oxadiazole, 1,2,4-oxadiazole, 3-chloromethyl-5-phenyl, 3-chloromethyl-5-phenyl-1,2,4-oxadiazol, 1,2,4-oxadiazole,3-chloromethyl-5-phenyl |
Formula Weight | 194.62 |
Percent Purity | 97% |
Chemical Name or Material | 3-(Chloromethyl)-5-phenyl-1, 2, 4-oxadiazole |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'