missing translation for 'onlineSavingsMsg'
Learn More

3-(Chloromethyl)-5-[4-(trifluoromethyl)phenyl]-1,2,4-oxadiazole, Tech., Thermo Scientific™

Product Code. 10120802
Change view
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
Product Code. Quantity Packaging unitSize
10120802 1 g Amber Glass Bottle 1g
1 options
This item is not returnable. View return policy

Product Code. 10120802

Brand: Thermo Scientific Maybridge SEW04003DA

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 175205-84-2
Packaging Amber Glass Bottle
Molecular Formula C10H6ClF3N2O
MDL Number MFCD00173952
Quantity 1 g
Synonym 3-chloromethyl-5-4-trifluoromethyl phenyl-1,2,4-oxadiazole, 3-chloromethyl-5-4-trifluoromethyl phenyl-1,2, 3-chloromethyl-5-4-trifluoromethyl-phenyl-1,2,4 oxadiazole, 3-chloromethyl-5-4-trifluoromethyl phenyl-1,2,4oxadiazole, 1,2,4-oxadiazole,3-chloromethyl-5-4-trifluoromethyl phenyl
InChI Key IOVWIIJODVBCRC-UHFFFAOYSA-N
SMILES FC(F)(F)C1=CC=C(C=C1)C1=NC(CCl)=NO1
IUPAC Name 3-(chloromethyl)-5-[4-(trifluoromethyl)phenyl]-1,2,4-oxadiazole
Molecular Weight (g/mol) 262.62
PubChem CID 2736624
Formula Weight 262.62
Percent Purity Tech.
Grade Technical
Chemical Name or Material 3-(Chloromethyl)-5-[4-(trifluoromethyl)phenyl]-1, 2, 4-oxadiazole
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.