missing translation for 'onlineSavingsMsg'
Learn More

3-(Chloromethyl)-5-[3-(trifluoromethyl)phenyl]-1,2,4-oxadiazole, ≥95%, Thermo Scientific™

Product Code. 10273272
Change view
Click to view available options
:
Amber Glass Bottle; 10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
10273272 Amber Glass Bottle; 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10273272 Supplier Thermo Scientific Maybridge Supplier No. SEW02034EA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

CAS 175205-63-7
CAS Min % 95.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Molecular Formula C10H6ClF3N2O
MDL Number MFCD00084969
Quantity 10 g
Synonym 3-chloromethyl-5-3-trifluoromethyl phenyl-1,2,4-oxadiazole, 1,2,4-oxadiazole, 3-chloromethyl-5-3-trifluoromethyl phenyl, 3-chloromethyl-5-3-trifluoromethyl-phenyl-1,2,4 oxadiazole, 3-chloromethyl-5-3-trifluoromethylphenyl-1,2,4 oxadiazole, pubchem12425, timtec-bb sbb005579, buttpark 48\08-26
InChI Key INANILNLXTUPHD-UHFFFAOYSA-N
SMILES FC(F)(F)C1=CC=CC(=C1)C1=NC(CCl)=NO1
IUPAC Name 3-(chloromethyl)-5-[3-(trifluoromethyl)phenyl]-1,2,4-oxadiazole
Molecular Weight (g/mol) 262.62
PubChem CID 2736622
Formula Weight 262.62
Percent Purity 95%
Chemical Name or Material 3-(Chloromethyl)-5-[3-(trifluoromethyl)phenyl]-1, 2, 4-oxadiazole
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.