missing translation for 'onlineSavingsMsg'
Learn More

3-(chloromethyl)-5-(3-thienyl)-1,2,4-oxadiazole, Thermo Scientific™

Product Code. 10702207
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10702207 5 g 5g
11360427 250 mg 250mg
10198312 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10702207 Supplier Thermo Scientific Maybridge Supplier No. SEW01730DE
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 184970-24-9
Molecular Formula C7H5ClN2OS
Molecular Weight (g/mol) 200.64
InChI Key KHJYOJVUQYWASE-UHFFFAOYSA-N
Synonym 3-chloromethyl-5-thiophen-3-yl-1,2,4-oxadiazole, 3-chloromethyl-5-3-thienyl-1,2,4-oxadiazole, 3-chloromethyl-5-thien-3-yl-1,2,4-oxadiazole, 1,2,4-oxadiazole,3-chloromethyl-5-3-thienyl, 3-3-chloromethyl-1,2,4-oxadiazol-5-yl thiophene
PubChem CID 2797043
IUPAC Name 3-(chloromethyl)-5-thiophen-3-yl-1,2,4-oxadiazole
SMILES C1=CSC=C1C2=NC(=NO2)CCl

Specifications

Assay Percent Range 97%
Packaging Amber glass bottle
Quantity 5 g
Formula Weight 200.65
Percent Purity 97%
Chemical Name or Material 3-(Chloromethyl)-5-(3-thienyl)-1, 2, 4-oxadiazole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.