missing translation for 'onlineSavingsMsg'
Learn More

3-(Chloromethyl)-5-(2-thienyl)-1,2,4-oxadiazole, ≥95%, Thermo Scientific™

Product Code. 10082214
Change view
Click to view available options
Quantity:
250 mg
1 g
10 g
Unit Size:
10g
1g
250mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10082214 10 g 10g
10613043 250 mg 250mg
10497632 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10082214 Supplier Thermo Scientific Maybridge Supplier No. SEW06262EA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 306936-06-1
Molecular Formula C7H5ClN2OS
Molecular Weight (g/mol) 200.64
MDL Number MFCD01571188
InChI Key YVYZVJRSESVBCM-UHFFFAOYSA-N
Synonym 3-chloromethyl-5-thiophen-2-yl-1,2,4-oxadiazole, 3-chloromethyl-5-thien-2-yl-1,2,4-oxadiazole, 3-chloromethyl-5-2-thienyl-1,2,4-oxadiazole, 3-chloromethyl-5-thiophen-2-yl-1,2,4 oxadiazole, 1,2,4-oxadiazole,3-chloromethyl-5-2-thienyl, 2-3-chloromethyl-1,2,4-oxadiazol-5-yl thiophene
PubChem CID 2798343
IUPAC Name 3-(chloromethyl)-5-thiophen-2-yl-1,2,4-oxadiazole
SMILES C1=CSC(=C1)C2=NC(=NO2)CCl

Specifications

CAS Min % 95.0
Assay Percent Range 95%
Quantity 10 g
Formula Weight 200.65
Percent Purity 95%
Chemical Name or Material 3-(Chloromethyl)-5-(2-thienyl)-1, 2, 4-oxadiazole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.