missing translation for 'onlineSavingsMsg'
Learn More

3-(Chloromethyl)-1-methyl-5-thien-2-yl-1H-pyrazole, ≥97%, Thermo Scientific™

Product Code. 10346904
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10346904 1 g Amber Glass Bottle 1g
10505364 250 mg Amber Glass Bottle 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10346904 Supplier Thermo Scientific Maybridge Supplier No. CC47124DA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 876316-61-9
Molecular Formula C9H9ClN2S
Molecular Weight (g/mol) 212.70
MDL Number MFCD08435883
InChI Key ODLZAEGCJDMPOT-UHFFFAOYSA-N
Synonym 3-chloromethyl-1-methyl-5-thien-2-yl-1h-pyrazole, 3-chloromethyl-1-methyl-5-thiophen-2-yl pyrazole, 1h-pyrazole,3-chloromethyl-1-methyl-5-2-thienyl, 3-chloromethyl-1-methyl-5-thiophen-2-yl-1h-pyrazole, 2-3-chloromethyl-1-methylpyrazol-5-yl thiophene
PubChem CID 18525807
IUPAC Name 3-(chloromethyl)-1-methyl-5-thiophen-2-ylpyrazole
SMILES CN1N=C(CCl)C=C1C1=CC=CS1

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 212.7
Percent Purity 97%
Chemical Name or Material 3-(Chloromethyl)-1-methyl-5-thien-2-yl-1H-pyrazole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.