missing translation for 'onlineSavingsMsg'
Learn More

3-(Chloromethyl)-1,2,4-oxadiazole, Technical Grade, Thermo Scientific™

£18.40 - £301.00

Chemical Identifiers

CAS 51791-12-9
Molecular Formula C3H3ClN2O
Molecular Weight (g/mol) 118.52
MDL Number MFCD00052097
InChI Key YSNKGJCEHOJIDK-UHFFFAOYSA-N
PubChem CID 2735765
IUPAC Name 3-(chloromethyl)-1,2,4-oxadiazole
SMILES ClCC1=NOC=N1
View More Specs

Products 3
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10283322
View Documents Promotion Details
Thermo Scientific Maybridge
SEW00285CB
250 mg Amber Glass Bottle
£18.40
250mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
10029491
View Documents Promotion Details
Thermo Scientific Maybridge
SEW00285DA
1 g Amber Glass Bottle
£54.15
1g
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
10041974
View Documents Promotion Details
Thermo Scientific Maybridge
SEW00285EA
10 g Amber Glass Bottle
£301.00
10g
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Chemical Identifiers

51791-12-9
118.52
YSNKGJCEHOJIDK-UHFFFAOYSA-N
3-(chloromethyl)-1,2,4-oxadiazole
C3H3ClN2O
MFCD00052097
2735765
ClCC1=NOC=N1

Specifications

51791-12-9
MFCD00052097
YSNKGJCEHOJIDK-UHFFFAOYSA-N
3-(chloromethyl)-1,2,4-oxadiazole
2735765
Tech.
3-(Chloromethyl)-1, 2, 4-oxadiazole
C3H3ClN2O
3-chloromethyl-1,2,4-oxadiazole, 1,2,4-oxadiazole,3-chloromethyl, chloromethyl 1,2,4-oxadiazole, 1,2,4-oxadiazole, 3-chloromethyl
ClCC1=NOC=N1
118.52
118.52
Technical
Videos
Safety and Handling

Safety and Handling

Danger

SDS
Documents

Documents

Product Certifications