missing translation for 'onlineSavingsMsg'
Learn More

3-Chlorobenzylamine, 98%

Product Code. 10613712 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10613712 5 g Glass bottle 5g
10225671 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10613712 Supplier Thermo Scientific Acros Supplier No. 297570050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 4152-90-3 | C7H8ClN | 141.6 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4152-90-3
Molecular Formula C7H8ClN
Molecular Weight (g/mol) 141.6
MDL Number MFCD00040752
InChI Key BJFPYGGTDAYECS-UHFFFAOYSA-N
Synonym 3-chlorobenzylamine, 3-chlorophenyl methanamine, benzenemethanamine, 3-chloro, 1-3-chlorophenyl methanamine, m-chlorobenzylamine, 3-chloro-benzylamine, m-chlorobenzyl amine, 3-chlorophenyl methylamine, chembl12957, c2a
PubChem CID 77802
IUPAC Name (3-chlorophenyl)methanamine
SMILES C1=CC(=CC(=C1)Cl)CN

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color Colorless to Yellow
Density 1.1500g/mL
Boiling Point 110.0°C to 112.0°C (17.0 mmHg)
Flash Point 98°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula ClC6H4CH2NH2
Refractive Index 1.5575 to 1.5605
Quantity 5 g
Specific Gravity 1.15
Formula Weight 141.6
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 3-Chlorobenzylamine
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.