missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Chlorobenzyl Chloride, TRC
Click to view available options
Quantity:
5 g
Unit Size:
5g
Specifications
Specifications
| Chemical Name or Material | 3-Chlorobenzyl Chloride |
| CAS | 620-20-2 |
| Molecular Formula | C7H6Cl2 |
| Quantity | 5 g |
| InChI Formula | InChI=1S/C7H6Cl2/c8-5-6-2-1-3-7(9)4-6/h1-4H,5H2 |
| SMILES | ClCc1cccc(Cl)c1 |
| IUPAC Name | 1-chloro-3-(chloromethyl)benzene |
| Molecular Weight (g/mol) | 161.03 |
| Formula Weight | 159.9847 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction