missing translation for 'onlineSavingsMsg'
Learn More

3-Chlorobenzyl chloride, 97%

Product Code. 11478873
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11478873 25 g 25g
11488873 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11478873 Supplier Thermo Scientific Alfa Aesar Supplier No. A11041.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 620-20-2 | C7H6Cl2 | 161.025 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 620-20-2
Molecular Formula C7H6Cl2
Molecular Weight (g/mol) 161.025
MDL Number MFCD00000905
InChI Key DDGRAFHHXYIQQR-UHFFFAOYSA-N
Synonym 3-chlorobenzyl chloride, 1-chloro-3-chloromethyl benzene, alpha,3-dichlorotoluene, m-chlorobenzyl chloride, benzene, 1-chloro-3-chloromethyl, 3-chlorobenzylchloride, m-chlorbenzyl chloride, 1-chloro-3-chloromethyl-benzene, toluene, m,.alpha.-dichloro, m,.alpha.-dichlorotoluene
PubChem CID 12103
IUPAC Name 1-chloro-3-(chloromethyl)benzene
SMILES C1=CC(=CC(=C1)Cl)CCl

Specifications

Density 1.27
Boiling Point 215°C to 216°C
Flash Point 98°C (208°F)
Odor Pungent
Refractive Index 1.556
Quantity 25 g
UN Number UN2235
Beilstein 742266
Formula Weight 161.03
Percent Purity 97%
Chemical Name or Material 3-Chlorobenzyl chloride
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.