missing translation for 'onlineSavingsMsg'
Learn More

3-chlorobenzonitrile, 99%

Product Code. 10249453 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 g
50 g
Packaging:
Glass bottle
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10249453 50 g Glass bottle 50g
10274130 10 g Glass bottle 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10249453 Supplier Thermo Scientific Acros Supplier No. 108680500
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 766-84-7 | C7H4ClN | 137.57 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 766-84-7
Molecular Formula C7H4ClN
Molecular Weight (g/mol) 137.57
MDL Number MFCD00001798
InChI Key WBUOVKBZJOIOAE-UHFFFAOYSA-N
Synonym m-chlorobenzonitrile, benzonitrile, 3-chloro, m-chloro benzonitrile, m-cyanochlorobenzene, benzonitrile, m-chloro, 3-chlorobenzenecarbonitrile, 5-chlorobenzonitrile, 3-chloro benzonitrile, 3-chloro-benzonitrile, pubchem3624
PubChem CID 13015
IUPAC Name 3-chlorobenzonitrile
SMILES ClC1=CC=CC(=C1)C#N

Specifications

Melting Point 38°C to 42°C
CAS Min % 98.5
Color Colorless to Yellow
CAS Max % 100.0
Density 1.1400g/mL
Quantity 50 g
Infrared Spectrum Authentic
Packaging Glass bottle
Physical Form Liquid
Assay Percent Range 98.5% min. (GC)
Linear Formula ClC6H4CN
Beilstein 09, 339
Boiling Point 94°C (11.0 mmHg)
Solubility Information Solubility in water: insoluble
Formula Weight 137.57
Percent Purity 99%
Chemical Name or Material 3-Chlorobenzonitrile
Show More Show Less
compliance-icons
Product Identifier
  • 3-Chlorobenzonitrile
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
  • H312-Harmful in contact with skin.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.