missing translation for 'onlineSavingsMsg'
Learn More

3-Chlorobenzoic acid, 99+%

Product Code. 10204090 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Plastic bottle
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10204090 100 g Plastic bottle 100g
10158652 25 g Plastic bottle 25g
10239453 500 g Plastic bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10204090 Supplier Thermo Scientific Acros Supplier No. 108661000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 535-80-8 | C7H5ClO2 | 156.57 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 535-80-8
MDL Number MFCD00002491
InChI Key LULAYUGMBFYYEX-UHFFFAOYSA-N
Synonym m-chlorobenzoic acid, benzoic acid, 3-chloro, benzoic acid, m-chloro, 3-chlorobenzoicacid, 3-chloro-benzoic acid, m-chlorobenzoate, acido m-clorobenzoico, unii-02uoj7064k, mcba, ccris 5992
PubChem CID 447
ChEBI CHEBI:49410
IUPAC Name 3-chlorobenzoic acid
SMILES C1=CC(=CC(=C1)Cl)C(=O)O

Specifications

CAS Min % 99.0
CAS Max % 100.0
Melting Point 154.0°C to 157.0°C
Color White to Beige
Flash Point 150°C
Infrared Spectrum Authentic
Assay Percent Range 99% min. (HPLC)
Packaging Plastic bottle
Molecular Formula C7H5ClO2
Linear Formula ClC6H4CO2H
Quantity 100 g
Beilstein 09, 337
Merck Index 15, 2124
Solubility Information Solubility in water: soluble in hot water. Other solubilities: freely soluble in alcohol and ether
Molecular Weight (g/mol) 156.57
Formula Weight 156.57
Percent Purity 99+%
Physical Form Crystalline Powder
Chemical Name or Material 3-Chlorobenzoic acid
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.