missing translation for 'onlineSavingsMsg'
Learn More

3-Chlorobenzo[b]thiophene-2-carboxylic acid, 97%

Product Code. 11334524
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11334524 1 g 1g
11344524 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11334524 Supplier Thermo Scientific Alfa Aesar Supplier No. L01257.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 21211-22-3 | C9H5ClO2S | 212.647 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 21211-22-3
Molecular Formula C9H5ClO2S
Molecular Weight (g/mol) 212.647
MDL Number MFCD00014579
InChI Key HJTMIYKPPPYDRJ-UHFFFAOYSA-N
Synonym 3-chlorobenzo b thiophene-2-carboxylic acid, 3-chloro-benzo b thiophene-2-carboxylic acid, 3-chloro-benzo b thiophene-2-carboxylicacid, benzo b thiophene-2-carboxylic acid, 3-chloro, 3-chlorobenzothiophene-2-carboxylic acid, pubchem18541, 3-chloro-benzo b thiophene carboxylic acid, acmc-1coc0, cbmicro_009255
PubChem CID 307964
IUPAC Name 3-chloro-1-benzothiophene-2-carboxylic acid
SMILES C1=CC=C2C(=C1)C(=C(S2)C(=O)O)Cl

Specifications

Melting Point 265°C to 267°C
Quantity 1 g
Beilstein 1425602
Formula Weight 212.65
Percent Purity 97%
Chemical Name or Material 3-Chlorobenzo[b]thiophene-2-carboxylic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.