missing translation for 'onlineSavingsMsg'
Learn More

3-Chlorobenzhydrazide, 98+%

Product Code. 11334444
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11334444 10 g 10g
11344444 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11334444 Supplier Thermo Scientific Alfa Aesar Supplier No. L01071.09

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 1673-47-8 | C7H7ClN2O | 170.596 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1673-47-8
Molecular Formula C7H7ClN2O
Molecular Weight (g/mol) 170.596
MDL Number MFCD00014757
InChI Key PHRDZSRVSVNQRN-UHFFFAOYSA-N
Synonym 3-chlorobenzhydrazide, 3-chlorobenzenecarbohydrazide, 3-chlorobenzoic hydrazide, benzoic acid, 3-chloro-, hydrazide, 3-chlorobenzoylhydrazine, 3-chlorobenzoic acid hydrazide, 3-chloro-benzoic acid hydrazide, benzoylhydrazine, m-chloro, 3-chlorobenzoic acid, hydrazide, 3-chlorobenzhyrazide
PubChem CID 74289
IUPAC Name 3-chlorobenzohydrazide
SMILES C1=CC(=CC(=C1)Cl)C(=O)NN

Specifications

Melting Point 156°C to 160°C
Quantity 10 g
Beilstein 973243
Formula Weight 170.6
Percent Purity ≥98%
Chemical Name or Material 3-Chlorobenzhydrazide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.