missing translation for 'onlineSavingsMsg'
Learn More

3-Chlorobenzenesulfonyl chloride, 98%

Product Code. 11454013
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11454013 5 g 5g
11444013 1 g 1g
11464013 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11454013 Supplier Thermo Scientific Alfa Aesar Supplier No. A10283.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 2888-06-4 | C6H4Cl2O2S | 211.06 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2888-06-4
Molecular Formula C6H4Cl2O2S
Molecular Weight (g/mol) 211.06
MDL Number MFCD00051697
InChI Key OINWZUJVEXUHCC-UHFFFAOYSA-N

Specifications

Melting Point ∼21°C
Density 1.499
Boiling Point 102°C to 104°C (1 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.569
Quantity 5 g
UN Number UN3265
Beilstein 2937139
Sensitivity Moisture sensitive
Synonym 3-methyl-1-phenyl-2-butanone, benzyl isopropyl ketone, isopropylbenzyl ketone, 2-butanone, 3-methyl-1-phenyl, 1-phenyl-3-methyl-2-butanone, unii-81804s3mkm, isopropyl benzyl ketone, isopropylbenzylketon, 5-nitrophenyl laurate, 2-methyl-4-phenyl-3-butanone
SMILES ClC1=CC=CC(=C1)S(Cl)(=O)=O
IUPAC Name 3-chlorobenzene-1-sulfonyl chloride
PubChem CID 76163
Formula Weight 211.07
Percent Purity 98%
Chemical Name or Material 3-Chlorobenzenesulfonyl chloride
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.