missing translation for 'onlineSavingsMsg'
Learn More

3-Chlorobenzeneboronic acid, 97%

Product Code. 11455897
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11455897 25 g 25g
11435897 1 g 1g
11445897 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11455897 Supplier Thermo Scientific Alfa Aesar Supplier No. B24444.14
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 63503-60-6 | C6H6BClO2 | 156.37 g/mol

It is employed as a reactant involved in 1,4-conjugate addition reactions with ethenesulfonamides to form arylethanesulfonamides, Suzuki-Miyaura reactions with dibromotrifluoromethylbenzene, cross-coupling reactions with diazoesters3 or potassium cyanate. It is involved in the synthesis of biarylketones and phthalides and synthesis of inhibitors including PDE4 inhibitors6 among others.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed as a reactant involved in 1,4-conjugate addition reactions with ethenesulfonamides to form arylethanesulfonamides, Suzuki-Miyaura reactions with dibromotrifluoromethylbenzene, cross-coupling reactions with diazoesters3 or potassium cyanate. It is involved in the synthesis of biarylketones and phthalides and synthesis of inhibitors including PDE4 inhibitors6 among others.

Solubility
Slightly soluble in water. soluble in ether, tetrahydrofuran,, dimethyl sulfoxide, dimethyl formamide, methanol.

Notes
Store at room temperature. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 63503-60-6
Molecular Formula C6H6BClO2
Molecular Weight (g/mol) 156.37
MDL Number MFCD00161354
InChI Key SDEAGACSNFSZCU-UHFFFAOYSA-N
Synonym 3-chlorophenyl boronic acid, 3-chlorobenzeneboronic acid, 3-chlorophenyl boranediol, 3-chlorophenylbornic acid, 3-chloro benzene boronic acid, m-chlorophenylboronic acid, 3-chloro-phenyl-boronic acid, boronic acid, 3-chlorophenyl, pubchem1785
PubChem CID 2734323
IUPAC Name (3-chlorophenyl)boronic acid
SMILES OB(O)C1=CC(Cl)=CC=C1

Specifications

Melting Point 180°C to 185°C
Quantity 25 g
Beilstein 2936343
Solubility Information Slightly soluble in water. soluble in ether,tetrahydrofuran,,dimethyl sulfoxide,dimethyl formamide,methanol.
Formula Weight 156.38
Percent Purity 97%
Chemical Name or Material 3-Chlorobenzeneboronic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.